5,6-Diaminouracil sulfate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5,6-Diamino-4-hydroxy-2(1H)-pyrimidinone sulphate, 5,6-Diamino-2,4-pyrimidinediol sulphate, 5,6-Diamino-2,4-dihydroxypyrimidine sulphate

43766-0100 43766-0500
TS43766-0100EA 177 USD
TS43766-0100 TS43766-0500
5,6-Diaminouracil sulfate 95%
5,6-Diaminouracil sulfate
Formula: C₄H₆N₄O₂.H₂SO₄
MW: 240.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00135845
CAS Number: 42965-55-9

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Elemental Analysis 21.93 to 27.40 % (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR