Ambrisentan

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-2-[(4,6-Dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropionic acid, (S)-2-((4,6-Dimethylpyrimidin-2-yl)oxy)-3-methoxy-3,3-diphenylpropanoic acid

46395-2500 463950010
TS46395-2500EA 160 USD
TS46395-2500 TS463950010
Ambrisentan
Ambrisentan
Formula: C₂₂H₂₂N₂O₄
MW: 378.42 g/mol
Melting Pt: >150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09842330
CAS Number: 177036-94-1

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR