Valnemulin hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

46283-0010 46283-2500
TS46283-0010EA 332 USD
TS46283-0010 TS46283-2500
Valnemulin hydrochloride
Valnemulin hydrochloride
Formula: C₃₁H₅₂N₂O₅S·HCl
Storage Temperature: Freezer
MDL Number: MFCD08458958
CAS Number: 133868-46-9
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Titration Argentometric 95.0 to 102.0 %
Water ≤4.0 %
HPLC ≥95.0 %
Specific optical rotation (Dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR