Propafenone hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-(2-Hydroxy-3-(propylamino)propoxy)phenyl)-3-phenylpropan-1-one hydrochloride, Propafenone HCl

46239-0010 46239-0050
TS46239-0010EA 61 USD
TS46239-0010 TS46239-0050
Propafenone hydrochloride
Propafenone hydrochloride
Formula: C₂₁H₂₇NO₃·HCl
MW: 377.9 g/mol
Melting Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00079243
CAS Number: 34183-22-7

Order Now

Specification Test Results

Appearance Colorless to white Powder
Infrared spectrum Conforms
Melting point 171 to 175 °C
Loss on drying ≤0.5 %
Solubility (5 % in methanol) Clear colorless solution
pH 5.0 to 6.2 (0.5 % in water)
Titration with HClO4 98 to 102 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR