Dexmedetomidine hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dex, (S)-Medetomidine hydrochloride, (+)-Medetomidine hydrochloride, (S)-4-[1-(2,3-Dimethylphenyl)ethyl]-1H-imidazole hydrochloride, (S)-4-(α,2,3-Trimethybenzyl)-1H-imidazole hydrochloride, Precedex, 5-[(1S)-1-(2,3-Dimethylphenyl)ethyl]-1H-imidazole hydrochloride

46543-2500 465430010
TS46543-2500EA 182 USD
TS46543-2500 TS465430010
Dexmedetomidine hydrochloride
Dexmedetomidine hydrochloride
Formula: C₁₃H₁₆N₂·HCl
MW: 236.74 g/mol
Melting Pt: 158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12912677
CAS Number: 145108-58-3

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 156 to 160 °C
HPLC ≥97.5 %
Specific optical rotation +48° to +54° (20 °C, 589 nm) (1% in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR