Dapagliflozin

Supplier: Thermo Scientific Chemicals
Warning

46462-0010 46462-0050
TS46462-0010EA 140 USD
TS46462-0010 TS46462-0050
Dapagliflozin
Dapagliflozin
Formula: C₂₁H₂₅ClO₆
Melting Pt: 65…70 °C
Storage Temperature: Freezer
MDL Number: MFCD13182359
CAS Number: 461432-26-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤2 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR