Tadalafil

Supplier: Thermo Scientific Chemicals

Synonyms: (6R,12aR)-6-(Benzo[d][1,3]dioxol-5-yl)-2-methyl-2,3,12,12a-tetrahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4(6H,7H)-dione

46194-0010 46194-0050
TS46194-0010EA 129 USD
TS46194-0010 TS46194-0050
Tadalafil
Tadalafil
Formula: C₂₂H₁₉N₃O₄
MW: 389.4 g/mol
Melting Pt: 296…303 °C
Storage Temperature: Freezer
MDL Number: MFCD07771966
CAS Number: 171596-29-5

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 3 h)
HPLC ≥96.0 %
Specific optical rotation +68° to +78° (20°C, 589 nm) (1% in CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR