Vinorelbine tartrate ≥98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl (3aR,3a1R,4R,5S,5aR,10bR)-4-acetoxy-3a-ethyl-9-((8R)-4-ethyl-8-(methoxycarbonyl)-1,3,6,7,8,9-hexahydro-2,6-methanoazecino[4,3-b]indol-8-yl)-5-hydroxy-8-methoxy-6-methyl-3a,3a1,4,5,5a,6,11,12-octahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylate bis((2R,3R)-2,3-dihydroxysuccinate), Vinorelbine ditartrate

46198-0250 46198-1000
TS46198-0250EA 165 USD
TS46198-0250 TS46198-1000
Vinorelbine tartrate ≥98%
Vinorelbine tartrate
Formula: C₅₃H₆₆N₄O₂₀
Storage Temperature: Freezer
MDL Number: MFCD03613607
CAS Number: 125317-39-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR