Amoxicillin 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2S,5R,6R)-6-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid

45514-0010 45514-0050 45514-0250
TS45514-0010EA 43 USD
TS45514-0010 TS45514-0050 TS45514-0250
Amoxicillin 96%
Amoxicillin
Formula: C₁₆H₁₉N₃O₅S
MW: 365.40 g/mol
Melting Pt: 330 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00056860
CAS Number: 26787-78-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water 10 to 15 %
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR