Rifapentine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2S,12Z,14E,16S,17S,18R,19R,20R,21S,22R,23S,24E)-3-(((4-Cyclopentyl-1-piperazinyl)imino)methyl)-21-(Acetyloxy)-5,6,9,17,19-pentahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-2,7-(epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione

46190-0250 46190-1000
TS46190-0250EA 104 USD
TS46190-0250 TS46190-1000
Rifapentine 95%
Rifapentine
Formula: C₄₇H₆₄N₄O₁₂
MW: 877.04 g/mol
Melting Pt: 179…180 °C
Storage Temperature: Freezer
MDL Number: MFCD00866806
CAS Number: 61379-65-5

Order Now

Specification Test Results

Appearance Red to dark red Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤8 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR