Sarpogrelate hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sarpogrelate HCl, 4-((1-(Dimethylamino)-3-(2-(3-methoxyphenethyl)phenoxy)propan-2-yl)oxy)-4-oxobutanoic acid hydrochloride

46191-0010 46191-2500
TS46191-0010EA 446 USD
TS46191-0010 TS46191-2500
Sarpogrelate hydrochloride 98%
Sarpogrelate hydrochloride
Formula: C₂₄H₃₁NO₆·HCl
MW: 465.97 g/mol
Melting Pt: 145…148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00887582
CAS Number: 135159-51-2
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 146 to 152 °C
Water ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR