Pyrantel pamoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Pyrantel embonate

46189-0050 46189-0250 46189-1000
TS46189-0050EA 59 USD
TS46189-0050 TS46189-0250 TS46189-1000
Pyrantel pamoate 98%
Pyrantel pamoate
Formula: C₁₁H₁₄N₂S·C₂₃H₁₆O₆
MW: 594.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00072037
CAS Number: 22204-24-6

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (60°C, 3 h) (vacuum)
Sulfated ash ≤0.5 %
Titration with HClO4 97 to 103 % (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR