Febuxostat 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(3-Cyano-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylic acid

46158-0010 46158-0050
TS46158-0010EA 131 USD
TS46158-0010 TS46158-0050
Febuxostat 98%
Febuxostat
Formula: C₁₆H₁₆N₂O₃S
MW: 316.37 g/mol
Melting Pt: 209 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00871598
CAS Number: 144060-53-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 207 to 211 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR