Procarbazine hydrochloride

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Isopropyl-4-[(2-methylhydrazino)methyl]benzamide, N-Isopropyl-4-((2-methylhydrazinyl)methyl)benzamide hydrochloride, Procarbazine HCl

46118-2500 461180010
TS46118-2500EA 178 USD
TS46118-2500 TS461180010
Procarbazine hydrochloride
Procarbazine hydrochloride
Formula: C₁₂H₁₉N₃O·HCl
MW: 257.76 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00072082
CAS Number: 366-70-1

Order Now

Specification Test Results

Appearance White to tan Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR