Meglumine antimoniate

Supplier: Thermo Scientific Chemicals
Warning

46114-0250 46114-1000
TS46114-0250EA 123 USD
TS46114-0250 TS46114-1000
Meglumine antimoniate
Meglumine antimoniate
Formula: C₇H₁₇NO₅·HO₃Sb
Storage Temperature: Ambient
MDL Number: MFCD20922853
CAS Number: 133-51-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to yellow Powder
Assay 30.0 to 34.0 % (Sb)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR