1,5-Anhydro-D-sorbitol 97%
Supplier: Thermo Scientific Chemicals
45991-0010
45991-2500
TS45991-0010EA
510
USD
TS45991-0010
TS45991-2500
1,5-Anhydro-D-sorbitol 97%
1,5-Anhydro-D-sorbitol
Formula:
C₆H₁₂O₅ Melting Pt: 141…144 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00067386 CAS Number: 154-58-5 |
Specification Test Results
Appearance | White Powder or crystalline powder |
Infrared spectrum | Conforms |
Melting point | 140 to 145 °C |
Elemental Analysis | ≥42.14 to ≤45.66 % Carbon |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...