Ciprofloxacin hydrochloride hydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride hydrate, 1-Cyclopropyl- 6-Fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid hydrochloride hydrate, Ciprofloxacin hydrochloride xhydrate

45688-0050 45688-0250
TS45688-0050EA 59 USD
TS45688-0050 TS45688-0250
Ciprofloxacin hydrochloride hydrate 98%
Ciprofloxacin hydrochloride hydrate
Formula: C₁₇H₁₈FN₃O₃·HCl·xH₂O
MW: 367.81 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00242856
CAS Number: 128074-72-6

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water 4.0 to 7.0 %
HPLC ≥97.5 %
Residue after ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR