N-Formyl-L-proline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-For-Pro-OH, (S)-1-Formylpyrrolidine-2-carboxylic acid, (S)-1-Formyl-2-pyrrolidinecarboxylic acid

45642-0010 45642-0050
TS45642-0010EA 114 USD
TS45642-0010 TS45642-0050
N-Formyl-L-proline 97%
N-Formyl-L-proline
Formula: C₆H₉NO₃
MW: 143.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037321
CAS Number: 13200-83-4

Order Now

Specification Test Results

Appearance White to off-white Powder or crystalline powder
Infrared spectrum Conforms
Elemental Analysis 9.39 to 10.17 % N
Specific optical rotation -115° to -130° (20°C, 589 nm) (c=0.8 in EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR