(S)-N-Boc-allylglycine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)pent-4-enoic acid, Boc-Allyl-Gly-OH, (S)-2-Boc-AMino-4-pentenoic acid, 4-Pentenoic acid, 2-[[(1,1-dimethylethoxy)carbonyl]amino]-, (2S)-

39010-0010 39010-0050 39010-5000
TS39010-0010EA 233 USD
TS39010-0010 TS39010-0050 TS39010-5000
(S)-N-Boc-allylglycine 95%
(S)-N-Boc-allylglycine
Formula: C₁₀H₁₇NO₄
MW: 215.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320851
CAS Number: 90600-20-7

Order Now

Specification Test Results

Appearance Colorless to yellow Viscous liquid or paste
Infrared spectrum Conforms
Specific optical rotation +9.0° to +13.0° (20°C, 589 nm) (c=1, methanol)
Titration with NaOH ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR