Kartogenin

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-((4-Phenylphenyl)carbamoyl)benzoic acid, 2-([1,1'-Biphenyl]-4-ylcarbamoyl)benzoic acid, 2-(N-(4-Phenylphenyl)carbamoyl)benzoic acid, N-Biphenyl-4-yl-phthalamic acid

46466-0010 464662500
TS46466-0010EA 454 USD
TS46466-0010 TS464662500
Kartogenin
Kartogenin
Formula: C₂₀H₁₅NO₃
Melting Pt: >280 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00560510
CAS Number: 4727-31-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to light yellow to beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR