(R)-N-Boc-α-ethylalanine 98%, 98% ee

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-D-isovaline, (R)-2-(tert-butoxycarbonylamino)-2-methylbutanoic acid, Boc-α-Et-D-Ala-OH, Boc-D-IsoVal-OH, (R)-2-((tert-Butoxycarbonyl)amino)-2-methylbutanoic acid

44106-1000 44106-5000
TS44106-1000EA 142 USD
TS44106-1000 TS44106-5000
(R)-N-Boc-α-ethylalanine 98%, 98% ee
(R)-N-Boc-alpha-ethylalanine
Formula: C₁₀H₁₉NO₄
MW: 217.265 g/mol
Melting Pt: 104…112 °C
Storage Temperature: Ambient
MDL Number: MFCD12963786
CAS Number: 123254-58-0

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Enantiomeric excess ≥98 % (HPLC)
Specific optical rotation -8° to -14° (20°C, 589 nm) (c=1 in CHCl3)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR