N,N-Dimethyl-L-phenylalanine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-2-(Dimethylamino)-3-phenylpropanoic acid, L-Phenylalanine, N,N-dimethyl-, Alanine, N,N-dimethyl-3-phenyl-, L-
44003-0010
44003-0050
TS44003-0010EA
29
USD
TS44003-0010
TS44003-0050
N,N-Dimethyl-L-phenylalanine 99%
N,N-Dimethyl-L-phenylalanine
Formula:
C₁₁H₁₅NO₂ MW: 193.246 g/mol Melting Pt: 225…227 °C Density: 0.095 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00082539 CAS Number: 17469-89-5 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Specific optical rotation | +73.0° to +77.0° (20°C, 589 nm) (c=1.3 in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...