L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43949-0010
43949-0050
TS43949-0010EA
159
USD
TS43949-0010
TS43949-0050
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride 95%
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride
Formula:
C₁₀H₁₄ClNO₄ Melting Pt: 94.2…94.3 °C Storage Temperature: Freezer |
MDL Number:
MFCD00038958 CAS Number: 1421-65-4 |
Specification Test Results
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
Water | ≤5.00 % |
Elemental Analysis | 5.32 to 5.99 % (N) On dry basis |
Specific optical rotation | +9.0° to +11.0° (20°C, 589 nm) (C=2 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...