Tetrabutylammonium hydroxide-30-hydrate 95+%

Supplier: Thermo Scientific Chemicals
Danger

43483-1000 43483-0250
TS43483-1000EA 172 USD
TS43483-1000 TS43483-0250
Tetrabutylammonium hydroxide-30-hydrate 95+%
Tetrabutylammonium hydroxide-30-hydrate
Formula: C₁₆H₉₇NO₃₁
Melting Pt: 27…30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149573
CAS Number: 147741-30-8
UN: 3263
ADR: 8,II

Order Now

Specification Test Results

Appearance Transparent white Crystals or crystalline powder
Melting point 26 to 30°C
Titration with HCl ≥95.0 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR