Axitinib

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: AG-013736, (E)-N-Methyl-2-((3-(2-(pyridin-2-yl)vinyl)-1H-indazol-6-yl)thio)benzamide

46380-0010 46380-2500
TS46380-0010EA 474 USD
TS46380-0010 TS46380-2500
Axitinib
Axitinib
Formula: C₂₂H₁₈N₄OS
MW: 386.5 g/mol
Melting Pt: 226…228 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09837898
CAS Number: 319460-85-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Almost white to yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR