Vindoline

Supplier: Thermo Scientific Chemicals
Warning

46227-0010 46227-0050
TS46227-0010EA 108 USD
TS46227-0010 TS46227-0050
Vindoline
Vindoline
Formula: C₂₅H₃₂N₂O₆
MW: 456.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD27991347
CAS Number: 2182-14-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR