L-Aspartic acid monosodium salt monohydrate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium (S)-3-amino-3-carboxypropanoate hydrate, Sodium L-aspartate monohydrate, L-Aspartic acid sodium salt monohydrate

45162-0250 45162-1000
TS45162-0250EA 26 USD
TS45162-0250 TS45162-1000
L-Aspartic acid monosodium salt monohydrate 97%
L-Aspartic acid monosodium salt monohydrate
Formula: C₄H₆NNaO₄·H₂O
MW: 173.11 g/mol (155.10 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00152960
CAS Number: 323194-76-9

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Water 8 to 12 %
Specific optical rotation +19.0° to +21.0° (20°C, 589 nm) (c=10, 2.4M HCl)
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR