Doxycycline monohydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

45889-0050 45889-0250
TS45889-0050EA 96 USD
TS45889-0050 TS45889-0250
Doxycycline monohydrate 97%
Doxycycline monohydrate
Formula: C₂₂H₂₄N₂O₈·H₂O
Melting Pt: 167…168 °C
Storage Temperature: Freezer
MDL Number: MFCD02682958
CAS Number: 17086-28-1

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Water 3.5 to 4.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR