DL-Histidine monohydrochloride monohydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-DL-His-OH.HCl.H2O, H-DL-His-OH·HCl·H₂O

41172-0250 41172-1000
TS41172-0250EA 67 USD
TS41172-0250 TS41172-1000
DL-Histidine monohydrochloride monohydrate 98%
DL-Histidine monohydrochloride monohydrate
Formula: C₆H₉N₃O₂·HCl·H₂O
MW: 209.63 g/mol (191.62 anhy)
Melting Pt: 253 °C
Storage Temperature: Ambient
MDL Number: MFCD00151028
CAS Number: 123333-71-1

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Water 7 to 10 %
Sulfated ash ≤0.2 %
Solubility light yellow solution
Titration with HClO4 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR