Fludrocortisone acetate

Supplier: Thermo Scientific Chemicals
Warning

46209-0010 462090050
TS46209-0010EA 138 USD
TS46209-0010 TS462090050
Fludrocortisone acetate
Fludrocortisone acetate
Formula: C₂₃H₃₁FO₆
Melting Pt: 224…226 °C
Storage Temperature: Freezer
MDL Number: MFCD00864160
CAS Number: 514-36-3

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C)
Solubility (5 % in acetone) Clear colorless to light yellow
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR