Ciclopirox olamine

Supplier: Thermo Scientific Chemicals
Danger

46344-0010 463440050
TS46344-0010EA 59 USD
TS46344-0010 TS463440050
Ciclopirox olamine
Ciclopirox olamine
Formula: C₁₂H₁₇NO₂·C₂H₇NO
Melting Pt: 143 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078997
CAS Number: 41621-49-2
UN: 3085
ADR: 5.1,I

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 21.9 to 23.7 % (2-Aminoethanol)
Titration with NaOH 74.2 to 80 % (Ciclopirox)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR