Cellulose acetate ∼ M.W. 100,000

Supplier: Thermo Scientific Chemicals

17778-5000 17778-0250
TS17778-5000EA 131 USD
TS17778-5000 TS17778-0250
Cellulose acetate ∼ M.W. 100,000
Cellulose acetate
Formula: C₂H₄O₂·(C₆H₁₀O₅)n
Melting Pt: 240 °C
Density: 1.31 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00081496
CAS Number: 9004-35-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Acetyl content 39.8 % (typical)
Hydroxyl content 3.6 % (typical)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR