α-D-Galactose-1-phosphate dipotassium salt pentahydrate 98%

Supplier: Thermo Scientific Chemicals

33703-0025 33703-0500 33703-5000
TS33703-0025EA 2211 USD
TS33703-0025 TS33703-0500 TS33703-5000
α-D-Galactose-1-phosphate dipotassium salt pentahydrate 98%
alpha-D-Galactose-1-phosphate dipotassium salt pentahydrate
Formula: C₆H₁₅K₂O₁₁P
Storage Temperature: Freezer
MDL Number: MFCD00150979
CAS Number: 19046-60-7

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤23 % (K.F.)
Solubility (5 % in water) Clear colorless
Specific optical rotation +81° to +84° (20°C, 589 nm) (c=2, H2O)
Titration with HClO4 ≥97.5 %
Phosphorus (P) ≤0.2 % (P inorganic)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR