L(+)-Isovaline monohydrate 99+%, (>99% ee)

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-(+)-2-Amino-2-methylbutanoic acid monohydrate

29005-0010 29005-2500
TS29005-0010EA 874 USD
TS29005-0010 TS29005-2500
L(+)-Isovaline monohydrate 99+%, (>99% ee)
L(+)-Isovaline monohydrate
Formula: C₅H₁₁NO₂.H₂O
Melting Pt: 305…308 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00167107
CAS Number: 1266371-20-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
TLC ≥99.0 %
Enantiomeric excess ≥99 % (GC)
Specific optical rotation +8.4° to +10.4° (25°C, 589 nm) (c=5, H2O)
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR