Trizma(R) maleate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Tris-maleate

26497-0250 26497-1000
TS26497-0250EA 40 USD
TS26497-0250 TS26497-1000
Trizma(R) maleate 98%
Trizma(R) maleate
Formula: C₈H₁₅NO₇
MW: 237.208 g/mol
Melting Pt: 104…106 °C
Storage Temperature: Ambient
CAS Number: 72200-76-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 104 to 106 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR