N-Acetyl-D-galactosamine 98%

Supplier: Thermo Scientific Chemicals

43492-0010 43492-0050
TS43492-0010EA 259 USD
TS43492-0010 TS43492-0050
N-Acetyl-D-galactosamine 98%
N-Acetyl-D-galactosamine
Formula: C₈H₁₅NO₆
Melting Pt: 158…166 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065372
CAS Number: 1811-31-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤7.0 %
Solubility (10 % in water) Clear colorless
HPLC ≥97.5 %
Specific optical rotation +80.0° to +90.0° (20°C, 589 nm) (c=1, water, after 5 hours)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR