Ouabain octahydrate 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Strophanthin G, 3-[(6-Deoxy-α-L-mannopyranosyl)oxy]-1,5,11α-14,19-penta-hydroxycard-20(22)-enolide
16173-0010
16173-0050
16173-0250
16173-2500
TS16173-0010EA
145
USD
TS16173-0010
TS16173-0050
TS16173-0250
TS16173-2500
Ouabain octahydrate 96%
Ouabain octahydrate
Formula:
C₂₉H₄₄O₁₂.8H₂O MW: 584.65 g/mol (anhy) Melting Pt: 260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149240 CAS Number: 11018-89-6 UN: 1544 ADR: 6.1,I |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | 18 % to 22 % (120°C, 2 hrs) |
Solubility | (1 % in water) Clear colorless |
HPLC | ≥95 % |
Specific optical rotation | -30° to -33° (20°C, 589 nm) (c=1, H2O) (on anhydrous substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...