Vinblastine sulfate 96.0-102.0%

Supplier: Thermo Scientific Chemicals
Danger

20345-0050 20345-0250
TS20345-0050EA 87 USD
TS20345-0050 TS20345-0250
Vinblastine sulfate 96.0-102.0%
Vinblastine sulfate
Formula: C₄₆H₆₀N₄O₁₃S
Melting Pt: 267 °C
Storage Temperature: Freezer
MDL Number: MFCD00082457
CAS Number: 143-67-9

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤15 %
HPLC 96.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR