Nω-Methyl-L-arginine monoacetate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Arg(Me)-OH.AcOH, N5-(Methylamidino)-L-ornithine acetate salt, Nomega-Monomethyl-L-arginine acetate, L-NMMA, Ng-Methyl-L-Arginine Acetate Salt, Nomega-Methyl-L-arginine acetate, N-ω-Monomethyl-L-arginine monoacetate, NG-Monomethyl-L-arginine acetate salt

32872-0050 32872-0250 32872-1000
TS32872-0050EA 38 USD
TS32872-0050 TS32872-0250 TS32872-1000
Nω-Methyl-L-arginine monoacetate 99%
Nω-Methyl-L-arginine monoacetate
Formula: C₇H₁₆N₄O₂·CH₃COOH
MW: 248.28 g/mol
Melting Pt: 180…190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00069311
CAS Number: 53308-83-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 180 to 190 °C
TLC ≥98.5 %
Specific optical rotation +9° to +13° (20 °C, 589 nm) (c=4, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR