L-Tyrosine disodium salt 98%

Supplier: Thermo Scientific Chemicals
Danger

42641-0250 42641-1000 42641-5000
TS42641-0250EA 78 USD
TS42641-0250 TS42641-1000 TS42641-5000
L-Tyrosine disodium salt 98%
L-Tyrosine disodium salt
Formula: C₁₈H₂₀N₂Na₂O₆
MW: 406.346 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
CAS Number: 69847-45-6

Order Now

Specification Test Results

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
Water ≤19.3 %
HPLC ≥97.5 %
pH 10.5 to 11.5 (1 % in water)
Specific optical rotation -9° to -14.5° (20°C, 589 nm) (c=4 in H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR