Boc-N-2-dimethylalanine 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: N-BOC-N,2-Dimethylalanine, Boc-N-Me-Aib-OH

37095-0010 37095-0050
TS37095-0010EA 104 USD
TS37095-0010 TS37095-0050
Boc-N-2-dimethylalanine 98+%
Boc-N-2-dimethylalanine
Formula: C₁₀H₁₉NO₄
MW: 217.265 g/mol
Melting Pt: 153…157 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862912
CAS Number: 146000-39-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 153 to 157 °C
Titration with NaOH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR