(1R,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid 95%, 98% ee
Supplier: Thermo Scientific Chemicals
Synonyms:
(1R,3R)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid
36239-0010
36239-5000
TS36239-0010EA
844
USD
TS36239-0010
TS36239-5000
(1R,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid 95%, 98% ee
(1R,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid
Formula:
C₁₁H₁₉NO₄ MW: 229.276 g/mol Melting Pt: 117.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD02259727 CAS Number: 489446-85-7 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
GC | ≥94 % |
Enantiomeric excess | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...