(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid 95%, 98% ee

Supplier: Thermo Scientific Chemicals

Synonyms: (1R,3S)-(+)-3-Aminocyclopentane-1-carboxylic acid, N-BOC protected, (1R,3S)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

36184-0010 36184-2500
TS36184-0010EA 370 USD
TS36184-0010 TS36184-2500
(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid 95%, 98% ee
(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid
Formula: C₁₁H₁₉NO₄
MW: 229.276 g/mol
Melting Pt: 96.1 °C
Storage Temperature: Ambient
MDL Number: MFCD01320858
CAS Number: 161660-94-2

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR