4-Borono-L-phenylalanine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-Phe(4-B)-OH, L-4-(2-Amino-2-carboxyethyl)benzeneboronic acid, (S)-2-Amino-3-(4-boronophenyl)propanoic acid

35893-2500 35893-1000
TS35893-2500EA 214 USD
TS35893-2500 TS35893-1000
4-Borono-L-phenylalanine 97%
4-Borono-L-phenylalanine
Formula: C₉H₁₂BNO₄
MW: 209.008 g/mol
Melting Pt: 283…293 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD01075172
CAS Number: 76410-58-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR