DL-2-Benzylserine 97%

Supplier: Thermo Scientific Chemicals
Warning

33978-0010 33978-0100
TS33978-0010EA 55 USD
TS33978-0010 TS33978-0100
DL-2-Benzylserine 97%
DL-2-Benzylserine
Formula: C₁₀H₁₃NO₃
MW: 195.218 g/mol
Melting Pt: 245…263 °C
Storage Temperature: Ambient
CAS Number: 4740-47-0

Order Now

Specification Test Results

Appearance Beige to light brown Crystalline powder or flakes
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR