Methylselenocysteine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Se-methylseleno-L-cysteine, (R)-2-Amino-3-(methylselanyl)propanoic acid, (R)-2-Amino-3-(methylseleno)propionic acid, Se-(Methyl)seleno-L-cysteine, H-3-(Me-Se)-Ala-OH

30064-0010 30064-0500 30064-2500
TS30064-0010EA 706 USD
TS30064-0010 TS30064-0500 TS30064-2500
Methylselenocysteine 98%
Methylselenocysteine
Formula: C₄H₉NO₂Se
Storage Temperature: Ambient
MDL Number: MFCD00800565
CAS Number: 26046-90-2
UN: 3283
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Specific optical rotation -13° to -15° (20°C, 589 nm) (c=1, water)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR