L-Methionine sulfoximine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2S)-2-Amino-4-(S-methylsulphonimidoyl)butanoic acid, H-Met(sulphoximine)-OH

22720-0010 22720-0050 22720-2500
TS22720-0010EA 302 USD
TS22720-0010 TS22720-0050 TS22720-2500
L-Methionine sulfoximine 98+%
L-Methionine sulfoximine
Formula: C₅H₁₂N₂O₃S
MW: 180.222 g/mol
Melting Pt: 210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002621
CAS Number: 15985-39-4

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Specific optical rotation +12° to +15° (20°C, 589 nm) (c=1, H2O)
Titration with HClO4 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR