2'-Deoxyguanosine-5'-monophosphate, sodium salt hydrate 99%

Supplier: Thermo Scientific Chemicals

22634-0010 22634-2500
TS22634-0010EA 126 USD
TS22634-0010 TS22634-2500
2'-Deoxyguanosine-5'-monophosphate, sodium salt hydrate 99%
2'-Deoxyguanosine-5'-monophosphate, sodium salt hydrate
Formula: C₁₀H₁₂N₅Na₂O₇P·xH₂O
Melting Pt: >245 °C
Storage Temperature: Freezer
MDL Number: MFCD00150780
CAS Number: 52558-16-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤20 % (K.F.)
Solubility (50 mg/ml in water) Clear colorless
HPLC ≥98.5 %
Ratio of Absorbances 0.25 to 0.28 (A290/A260) c=1, water

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR