Cellulose triacetate
Supplier: Thermo Scientific Chemicals
17782-2500
17782-5000
TS17782-2500EA
100
USD
TS17782-2500
TS17782-5000
Cellulose triacetate
Cellulose triacetate
Formula:
C₄₀H₅₄O₂₇ MW: 966.845 g/mol Storage Temperature: Ambient |
CAS Number:
9012-09-3 |
Specification Test Results
Appearance | White to light yellow Pellets or rods |
Infrared spectrum | Conforms |
Acetyl content | 43 to 44 wt % |
Titration Argentometric | ≥95 % (on dry substance) |
Water | ≤10 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...