Cefaclor 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (6R,7R)-7-((R)-2-Amino-2-phenylacetamido)-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

45526-0010 45526-0050
TS45526-0010EA 76 USD
TS45526-0010 TS45526-0050
Cefaclor 96%
Cefaclor
Formula: C₁₅H₁₄ClN₃O₄S
MW: 385.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151471
CAS Number: 53994-73-3

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water 3.0 to 9.0 %
HPLC 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR